Universal pH Indicator Paper
Product Image
Fisherbrand
Universal pH Indicator Paper
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH Range
FKS117835540 to 10
FKS117035645 to 9
Catalog number FKS11783554
Price (SGD)
18.00
EA
Add to cart
pH Range:
0 to 10
Price (SGD)
18.00
EA
Add to cart
  • Filter paper soaked with one or several colored indicators of pH
  • Economic range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
pH Range0 to 10
FormatRoll
Quantity1 Roll
TypeGrade 911
Length (Metric)5 m
Width (Metric)10 mm
Unit SizeEA