Traceable™ Digital Calipers
Traceable™ Digital Calipers
Actual product may vary
Fisherbrand
Traceable™ Digital Calipers
Ideal for measuring the outside diameter of stoppers, the inside diameter of tubing, and the depth of vials. Set jaws closed, or read differential measurements with the touch of a button.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeWeight (English)
FKS117832170 to 150 mm5 oz.
CTL06-664-160 to 200 mm6 oz.
CTL14-648-170 to 150 mm5 oz.
Catalog number FKS11783217
Price (SGD)
488.00
EA
Add to cart
Size:
0 to 150 mm
Weight (English):
5 oz.
Price (SGD)
488.00
EA
Add to cart
Measure the outside diameter of stoppers, the inside diameter of tubing, and the depth of vials. Read differential measurements with the touch of a button.
  • 0.33 in. high digital display
  • Durable hardened steel
  • Resolution: 0.0005 in. 
  • Accuracy: ±0.001 in. 
9 in. Model
  • Range: 0 to 6 in. 
  • Dimensions: 9 x 3 in. (22.9 x 7.6 cm)
  • Weight: 5 oz. (141.7g)
11 in. Model
  • Range: 0 to 8 in. 
  • Dimensions: 11 x 3 in. (27.9 x 7.6 cm)
  • Weight: 6 oz. (170g)
Specifications
Product TypeCaliper, Traceable Digital
Accuracy±0.001 in.
For Use With (Application)Ideal for measuring the outside diameter of stoppers, the inside diameter of tubing, and the depth of vials
Includes1.5V Button cell 357 battery, carrying case, Traceable™ certificate
Scale TypeCalibrated in inches
Size0 to 150 mm
Weight (English)5 oz.
Weight (Metric)0.14 kg
MaterialStainless Steel
DescriptionLab tools resolution: 0.01mm
Unit SizeEA