Stainless Steel Griffin-Style Beaker
Stainless Steel Griffin-Style Beaker
Actual product may vary
Fisherbrand
Stainless Steel Griffin-Style Beaker
Use these beakers for a variety of applications. Cole-Parmer™ Stainless Steel Griffin-Style Beakers are ideal for general stirring or measuring of solutions. Beakers feature an easy-pour rim for fewer spills.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11782315600 mL
FKS11762315125 mL
Catalog number FKS11782315
Price (SGD)
487.00
EA
Add to cart
Capacity (Metric):
600 mL
Price (SGD)
487.00
EA
Add to cart
Use these beakers for a variety of applications. Cole-ParmerTM Stainless Steel Griffin-Style Beakers are ideal for general stirring or measuring of solutions. Beakers feature an easy-pour rim for fewer spills.
Griffin-style beakers with pouring spout have full capacity mark near the top to assure accurate measuring.
  • Durable, economical mixing capabilities
  • Made of medium gauge 304 Stainless Steel
  • Non-corrosive
  • Easy to clean
Specifications
Diameter (Metric) Outer85mm
Capacity (Metric)600 mL
MaterialStainless Steel
Height (Metric)120mm
Temperature (Metric) Max. Operating426.7°C
TypeBeaker