Traceable™ Humidity-On-A-Card™ Humidity Monitor
Traceable™ Humidity-On-A-Card™ Humidity Monitor
Actual product may vary
Fisherbrand
Traceable™ Humidity-On-A-Card™ Humidity Monitor
Traceable™ Humidity-On-A-Card™ Humidity Monitor ideal for use with bottles, chemicals, plastic bags, electronics, desiccators and storage containers.
Have Questions?
Catalog number FKS11782146
Price (SGD)
-
Traceable™ Humidity-On-A-Card™ Humidity Monitor ideal for use with bottles, chemicals, plastic bags, electronics, desiccators and storage containers.
  • Cards record both the current humidity and the maximum humidity attained using color-change reactions
  • Current humidity scale ranges from 0 to 100% RH in 10% increments
  • Color changes from blue to bright pink as the humidity levels rise and fall
  • Accuracy is ±5% RH at 75°F (24°C)
  • Maximum humidity scale uses a nonreversible color reaction to record the highest humidity level to which the card has been exposed (over a 24-hour period)
  • Range is 50 to 90% RH in 10% increments
  • Accuracy: ±5% RH at 75°F (24°C)
  • Each card is individually sealed in a clear moisture-barrier bag with a Humidity Sponge™ desiccant to keep the card dry and to provide for an extended shelf life
Specifications
TypeHumidity Monitor
Humidity Range0 to 100%
Accuracy±5% RH at 75°F (24°C)
Temperature Range (Metric)24°C
Increments0.1
Temperature Range (English)75°F
For Use With (Application)Monitor humidity levels in hard-to-reach or out-of-the-way areas over extended time periods, record both the current humidity and the maximum humidity attained using color-change reactions
IncludesIndividually serial-numbered ISO 17025 certificate
Certifications/ComplianceNIST
Unit SizeEA