Water-Resistant, Shock-Resistant Stopwatch
Water-Resistant, Shock-Resistant Stopwatch
Actual product may vary
Fisherbrand
Water-Resistant, Shock-Resistant Stopwatch
NIST traceability assures quality and accuracy. Fisherbrand Water-Resistant, Shock-Resistant Stopwatch is a general-purpose, quartz LCD digital stopwatch with O-ring-sealed ABS plastic case and is ideal for rough use.
Have Questions?
Catalog number FKS11779825
Price (SGD)
78.00
EA
Add to cart
Price (SGD)
78.00
EA
Add to cart
  • Times to 24 hours with a resolution of 0.01 of a second
  • Features single-action, time-out, cumulative split and continuous timing
  • Displays time-of-day in date and the day of the week
Specifications
Product TypeWater and Shock-resistant Stopwatch
Time Range24 hr.
AlarmsAudible
Accuracy0.0001
Resolution0.01 sec.
DisplayLCD
MaterialABS Plastic
Dimensions (L x W x H)2.12 x 2.37 x 0.5 in. (5.4 x 6 x 1.27 cm)
Length (English)2.12 in.
Length (Metric)5.4 cm
Width (English)2.37 in.
Width (Metric)6 cm
Height (English)0.5 in.
Height (Metric)1.27 cm
Weight (English)1 oz.
Weight (Metric)0.028 kg
Includes2 Year battery, lanyard and instructions
For Use With (Application)Ideal for lab or plant use
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEA