Triangular Tripod Stand
Triangular Tripod Stand
Actual product may vary
Triangular Tripod Stand
Triangular Tripod Stand
Triangular Tripod Stand
Fisherbrand
Triangular Tripod Stand
Designed to cope with the demands of a modern laboratory.
Have Questions?
Catalog NumberLength (Metric)
FKS11779806125 mm side length
Catalog number FKS11779806
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
125 mm side length
  • Manufactured in zinc-plated steel
  • Legs of 6mm in diameter
  • Legs riveted in place and bent outwards for stability
Specifications
MaterialStainless Steel
Height (Metric)210 mm
Length (Metric)125 mm side length
Product TypeTripod Stand
ShapeTriangular
FinishZinc Plated
Unit SizeEA