Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani)
Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani)
Actual product may vary
Fisherbrand
Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani)
Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging Type
FKS11778902500 gBottle
FISDF0446-07-52000 gPail
FISDF0446-17-3500 gBottle
Catalog number FKS11778902
Price (SGD)
337.00
EA
Add to cart
Quantity:
500 g
Packaging Type:
Bottle
Price (SGD)
337.00
EA
Add to cart
  • Contains 10g/L NaCl
Specifications
Certifications/ComplianceCCAM
FormDehydrated
Media TypeLB Broth, Miller
Product TypeMicrobiology Media
FormulationTryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application)Propagation and maintenance of Escherichia coli for molecular biology
Packaging TypeBottle
Quantity500 g
Shelf Life1825 Days
Unit SizeEA
Contents & Storage
Refrigerate