Screw-Top Bottle, Square, Amber, Soda-Lime Glass, Wide mouth, Short form DIN thread
Screw-Top Bottle, Square, Amber, Soda-Lime Glass, Wide mouth, Short form DIN thread
Actual product may vary
Fisherbrand
Screw-Top Bottle, Square, Amber, Soda-Lime Glass, Wide mouth, Short form DIN thread
Square amber soda lime glass bottles, wide mouth.
Have Questions?
Catalog NumberCapacity (Metric)
FKS11777991100 mL
Catalog number FKS11777991
Price (SGD)
123.00
PK
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
123.00
PK
Add to cart
Square amber soda lime (Type 3) glass bottles, wide mouth and short form DIN thread.
  • Amber glass provides UV protection
  • Square shape for efficient packing
Specifications
Capacity (Metric)100 mL
For Use With (Application)Storage of light-sensitive materials; Transport and shipping of light-sensitive materials
MaterialSoda Lime Glass
Quantity10/Pk.
Thread Size (Metric)GL 32
AutoclavableNot Autoclavable
Diameter (Metric)49 mm
Unit SizePK