Traceable™ Digital Carbon Fiber Calipers
Traceable™ Digital Carbon Fiber Calipers
Actual product may vary
Traceable™ Digital Carbon Fiber Calipers
Traceable™ Digital Carbon Fiber Calipers
Fisherbrand
Traceable™ Digital Carbon Fiber Calipers
Switch between inches and millimeters easily
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWeight (English)
FKS117771051 oz.
CTL15-077-9571 oz.
CTL15-077-9582 oz.
Catalog number FKS11777105
Price (SGD)
161.00
EA
Add to cart
Weight (English):
1 oz.
Price (SGD)
161.00
EA
Add to cart
Switch between inches and millimeters easily
For measuring the outside or inside diameter. Ergonomically designed. No lubrication is ever required on the ultra-smooth slide bar.
  • Large digital display: 0.3 in.H
  • Made of strong composite carbon fiber
  • Greater-than or less-than results without calculations
  • Zero button instantly resets unit at any position
  • Runs on the supplied battery for 10,000 hours
  • Resolution: 0.01 in. and 0.1mm
  • Accuracy: ±0.01 in. and ±0.2mm
4 in. version
  • Range: 0 to 4 in. (0 to 100mm)
  • Dimensions: 7 x 2.5 in.
  • Weight: 1 oz.
6 in. version
  • Range: 0 to 6 in. (0 to 150mm)
  • Dimensions: 9.5 x 3 in.
  • Weight: 2 oz.
Specifications
Product TypeCaliper, Digital
For Use With (Application)Measures both inside and outside dimensions
Weight (English)1 oz.
Weight (Metric)0.25 kg
MaterialCarbon Fiber
Unit SizeEA