HisTrap™ HP Prepacked Columns
HisTrap™ HP Prepacked Columns
Actual product may vary
HisTrap™ HP Prepacked Columns
HisTrap™ HP Prepacked Columns
Fisherbrand
HisTrap™ HP Prepacked Columns
Purify histidine-tagged proteins directly from homogenized, unclarified cells. Cytiva HisTrap™ HP Prepacked Columns are prepacked with Ni Sepharose™ High Performance and designed for simple, high-resolution purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
Have Questions?
Catalog NumberQuantityDiameter (Metric) Inner
FKS117732095 x 5 mL16 mm
Catalog number FKS11773209
Price (SGD)
-
Quantity:
5 x 5 mL
Diameter (Metric) Inner:
16 mm
  • Prepacked with Ni Sepharose™ High Performance medium
  • Negligible nickel ion leakage
  • Pressure Max: 3 bar
  • Column material: polypropylene
  • 1 and 5mL columns
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Manual operation with syringe, pump or chromatography system
Media
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 12
1mL columns
  • Bed Dimensions: 7 x 25mm
  • Flow velocity: 4mL/min.
5mL columns
  • Bed Dimensions: 16 x 25mm
  • Flow velocity: 5mL/min.
Specifications
Bed Volume5 mL
Diameter (English) Inner0.6 in.
Diameter (Metric) Inner16 mm
DescriptionHisTrap HP are nickel-charged IMAC columns for high resolution his-tagged protein purification with high binding capacity for maximized recovery.
Flow Rate4 mL/min. (1 mL), 5 mL/min. (5 mL)
MaterialPolypropylene
pH Range3 to 12
Support Type6% Highly Cross-linked Agarose
Max. Operating Pressure5 bar (0.5 MPa, 70 psi)
Quantity5 x 5 mL
TypePre-packed Column
Unit SizePK
Contents & Storage
4°C to 30°C