Check Order Status
Biohazard Pouch
Actual product may vary
Fisherbrand
Biohazard Pouch
Double-pocketed, self-adhesive biohazard bags printed with a prominent biohazard logo. Gosselin™ Biohazard Pouch is designed for decontaminating laboratory waste via autoclaving.
Have Questions?
Catalog Number
Length (Metric)
FKS11771605
170 mm
Catalog number
FKS11771605
Price (SGD)
1,832.00
PK
Add to cart
Length (Metric):
170 mm
Price (SGD)
1,832.00
PK
Add to cart
Durable polypropylene construction
Conical shape
Flat bottom
Red screw-cap closure
Specifications
Closure Type
Self-Adhesive Closure
Height (Metric)
270 mm
Length (Metric)
170 mm
Quantity
2000/Cs.
Thickness
40 μm