Round-Base 3.5mL Test Tubes
Round-Base 3.5mL Test Tubes
Actual product may vary
Round-Base 3.5mL Test Tubes
Providing high resistance to centrifugation up to 3000g with a suitable adaptor in a balanced rotor. Gosselin™ Round-Base 3.5mL Test Tubes are available in either polystyrene or polypropylene.
Have Questions?
Catalog NumberMaterialSterilityClosure Type
FKS11769402PolystyreneNon-sterileNot Included
Catalog number FKS11769402
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
Polystyrene
Sterility:
Non-sterile
Closure Type:
Not Included
  • For laboratory use
  • All tubes are CE + IVD marked
  • Available with or without closure
  • Compatible with automation systems
  • Available in sterile or non-sterile (sterility provided by electron beam treatment at 10kGy)
Specifications
Certifications/ComplianceEC N°1935/2004
ColorNatural
Capacity (Metric)3.5 mL
MaterialPolystyrene
For Use WithClinical, Dairy, Environment, Industries, Microbiology
Packaging500BG
Quantity3000 Carton
RimmedNo
SterilityNon-sterile
Diameter (Metric)11.8 mm
Closure TypeNot Included
Product TypeTest Tube
Unit SizePK