Sterilin™ Clear Triple Vent Petri Dishes
Sterilin™ Clear Triple Vent Petri Dishes
Actual product may vary
Fisherbrand
Sterilin™ Clear Triple Vent Petri Dishes
Culture microorganisms on solid media using Thermo Scientific Sterilin Standard 90mm Petri Dishes. These shallow dishes maximize incubator space and feature a mirror finish that ensures high optical quality.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1176925290 mm
Catalog number FKS11769252
Price (SGD)
547.00
PK
Add to cart
Diameter (Metric):
90 mm
Price (SGD)
547.00
PK
Add to cart
  • Manufactured from non-cytotoxic virgin polystyrene
  • Ideal for use in automatic plate pourers
  • For in vitro use only
  • Triple vented—aids gaseous exchange; ideally suited for short-term work
Specifications
Certifications/ComplianceCE, 98/79/EC, BS611 part 2
ColorClear
MaterialPolystyrene
ShapeRound
SterilitySterile
TypePetri Dish
For Use With (Application)For use in automatic plate pourers, for in vitro use only
Quantity500 Case
Height (Metric)16.2 mm
Diameter (Metric)90 mm
Unit SizePK