Grade 122 Cellulose General Purpose Filter Paper
Grade 122 Cellulose General Purpose Filter Paper
Actual product may vary
Fisherbrand
Grade 122 Cellulose General Purpose Filter Paper
Very fast filtration velocity. Fisherbrand™ Grade 122 Cellulose General Purpose Filter Paper is used for all general works in laboratories.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11768742250 mm
Catalog number FKS11768742
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
250 mm
  • Very fast filtration velocity
  • Used for general works, hospitals, research laboratories
Applications:
  • Chemical industries, pharmaceutical, cosmetic, food and beverage
  • Specifications
    FormatDisc
    MaterialCellulose
    Particle Retention17 to 30 μm
    Pore Size17 to 30 μm
    For Use With (Application)For all general works in laboratories
    Grade122
    Quantity100/Pk.
    Thickness (Metric)0.17 mm
    Diameter (Metric)250 mm
    Diameter (Metric) Disc250 mm
    Unit SizePK