Disposable PD-10 Desalting Columns
Disposable PD-10 Desalting Columns
Actual product may vary
Fisherbrand
Disposable PD-10 Desalting Columns
Use for desalting and buffer exchange, as well as removal of low-molecular weight compounds. Cytiva Life Sciences™ Disposable PD-10 Desalting Columns provide fast, high-recovery.
Have Questions?
Catalog NumberQuantity
FKS1176848830 Pk.
Catalog number FKS11768488
Price (SGD)
-
Quantity:
30 Pk.
  • Packed with Sephadex™ G-25 medium
  • Designed for ease of use and equilibration
  • Prepare samples in range of 1.0 to 2.5mL by gravity flow or 1.75 to 2.5mL by centrifugation
  • Desalt multiple samples in parallel with desalting capacity >90%
  • Sample recovery in range of 70 to >95%
  • Includes adapters for centrifugation
Specifications
Product TypeDesalting Column, Sephadex G-25 Medium Packing
Particle Size>50 μm (dry), 38 to 235 μm (wet)
Diameter (English) Inner Column0.59 in.
For Use With (Application)Designed for ease of use and equilibration
Volume (Metric) Bed8.3 mL
Diameter (Metric) Inner Column1.5 cm
Quantity30 Pk.
Unit SizePK
Contents & Storage
4°C to 30°C