Aluminum Turbine Flowmeter
Aluminum Turbine Flowmeter
Actual product may vary
Fisherbrand
Aluminum Turbine Flowmeter
Have Questions?
Catalog NumberFlow Rate Range
FKS117673283 to 50 GPM
Catalog number FKS11767328
Price (SGD)
4,307.00
EA
Add to cart
Flow Rate Range:
3 to 50 GPM
Price (SGD)
4,307.00
EA
Add to cart
Specifications
TypeTurbine Flowmeter
Accuracy±1.5% (Reading)
Float MaterialAluminum
Pressure Rating300 psig
Connections1 in. NPT (F)
Flow Rate Range3 to 50 GPM
For Use With (Application)Low-cost solution to light-duty process applications
Height (Metric)64 mm
Power Supply DetailsTwo 3 V Lithium Batteries
Repeatability±0.2%
Width (Metric)110 mm
Temperature (Metric)60°C
Weight (Metric)600 g
Unit SizeEA