G2 Heavy-Duty Turbine Flowmeter/Totalizer
G2 Heavy-Duty Turbine Flowmeter/Totalizer
Actual product may vary
Fisherbrand
G2 Heavy-Duty Turbine Flowmeter/Totalizer
Designed to handle most basic industrial flow metering applications. Cole-Parmer™ G2 Heavy-Duty Turbine Flowmeter/Totalizer features two totalizers allow for process or period-specific volume monitoring. Sanitary units are perfect for micro-brewing and wine making processes.
Have Questions?
Catalog number FKS11767238
Price (SGD)
6,503.00
EA
Add to cart
Price (SGD)
6,503.00
EA
Add to cart
  • Battery-powered to eliminate the need for expensive wiring
  • Displays flow rate and accumulated flow in GPM or LPM
  • NEMA enclosure is suitable for indoor/outdoor use in dusty, dirty and wet environments
  • Compact yet robust design ideal for use in manufacturing, pilot-plant or quality control areas
  • All flowmeters measure linearly across a range and record a cumulative volume plus a resettable volume
  • Factory-calibrated using water but can store 2 application-specific K-factors determined in-process
  • To conserve power, the six-digit display automatically turns on with flow and then turns off four minutes after flow ceases
  • Wetted components are 316 SS or aluminum housing, PVDF rotor and supports, tungsten carbide shaft, 316 SS retainers, and ceramic bearings
  • The internal parts are simple to replace for easy maintenance
  • Meters operate on two lithium batteries
  • Remote installation of the flowmeter body is possible using the optional remote installation kit (order separately).
  • Replacement battery kits, output modules and calibration containers available
Specifications
Accuracy±2%
Airflow Rate1 to 10 GPM
Certifications/ComplianceEnclosure is NEMA 4 for Class I, Division I environments
For Use With (Application)Basic industrial flow metering applications
Internal Part MaterialPVDF rotor and supports, Tungsten Carbide shaft, Stainless Steel (316) retainers, and ceramic bearings
Repeatability±0.1%
Temperature (English)Up to 60°F
Temperature (Metric)140°C max.
Unit SizeEA