Endecotts™ Stainless Steel Consistometer
Endecotts™ Stainless Steel Consistometer
Actual product may vary
Fisherbrand
Endecotts™ Stainless Steel Consistometer
Use this consistometer to determine the consistency of viscous materials by measuring the distance that the material flows under its own weight in a given time interval. Cole-Parmer™ Endecotts™ Stainless Steel Consistometer checks against consistency, viscosity and flow rate standards.
Have Questions?
Catalog NumberVolume (Metric)
FKS1176537975 mL
Catalog number FKS11765379
Price (SGD)
4,823.00
EA
Add to cart
Volume (Metric):
75 mL
Price (SGD)
4,823.00
EA
Add to cart
Use this consistometer to determine the consistency of viscous materials by measuring the distance that the material flows under its own weight in a given time interval. Cole-Parmer™ Endecotts™ Stainless Steel Consistometer checks against consistency, viscosity and flow rate standards.
  • Verify the consistency, viscosity and flow rates of fluids
  • Meets military specification R-81294B for paint manufacturing
  • Ideal for predetermining product formulas to standardize production lots
  • Consistometer requires only 75mL of sample for measurement
  • Engraved graduations in 0.5cm divisions ensure accurate results
  • Spring-loaded gate prevents premature sample flow
  • Stainless steel construction prevents corrosion
Specifications
Product TypeConsistometer
MaterialStainless Steel
Height (Metric)14 cm
Length (Metric)35.6 cm
Width (Metric)8.9 cm
Volume (Metric)75 mL
Unit SizeEA