Sample at specific depths between 3 and 600 feet. Cole-Parmer™ Wildco 1510-C20 Kemmerer Water Sampler features an acrylic bottle with silicone end seals.
Have Questions?
Catalog Number
Capacity (Metric)
FKS11764499
2.2 L
Catalog number FKS11764499
Price (SGD)
5,769.00
EA
Add to cart
Capacity (Metric):
2.2 L
Price (SGD)
5,769.00
EA
Add to cart
Messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle
Order messenger and line separately
Includes a plastic carrying case
Specifications
Product TypeWater Sampler
IncludesPlastic carrying case
Closure MaterialSilicone
Tube MaterialAcrylic
Capacity (Metric)2.2 L
StyleKemmerer Water Sampler
For Use With (Application)Water sampling at depths of 3 to 600 ft.