Wide-mouth Soda Lime Glass Packer Bottles
Product Image
Fisherbrand
Wide-mouth Soda Lime Glass Packer Bottles
Designed with a wide mouth for easy accessibility, these bottles are ideal storage containers. Fisherbrand™ Wide-mouth Soda Lime Glass Packer Bottles are clear and autoclavable.
Have Questions?
Catalog NumberCapacity (Metric)
FKS11764329500 mL
Catalog number FKS11764329
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
  • Clear soda lime glass
  • Autoclavable (lids are NOT)
  • Polypropylene cap with polyvinyl liner
  • Ideal for storage of dry powders
  • Wide mouth for easy accessibility
Specifications
Capacity (Metric)500 mL
ColorClear
Diameter (Metric) Inner Neck50.1mm
MaterialSoda Lime Glass
Quantity24/PK.
Thread Size (Metric)Style: 63/400
Closure ColorWhite
Liner MaterialPolyvinyl
Closure IncludedYes
MouthWide
Height (Metric)133 mm
Closure MaterialPolypropylene
Closure Size63-400 mm
Diameter (Metric)79 mm
Closure TypeScrew Cap
Unit SizePK