Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam/PTFE Cap
Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam/PTFE Cap
Actual product may vary
Fisher Catalogue
Amber Wide Mouth Packer Bottles with White Polypropylene PE Foam/PTFE Cap
Feature a wide opening, making them ideal general purpose bottles for solids. Fisherbrand™ Amber Wide-Mouth Packers with PTFE-Faced PE-Lined Caps are the classic vitamin bottles, and are often used in pharmaceutical, chemical, and environmental applications. The amber glass filters out ultraviolet wavelengths to protect light-sensitive solids.
Have Questions?
Catalog NumberCapacity (Metric)Closure SizeQuantity
FKS1176424960 mL33-400 mm24/Cs.
Catalog number FKS11764249
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
60 mL
Closure Size:
33-400 mm
Quantity:
24/Cs.
Amber Glass Wide Mouth Packer Bottles feature a wide opening for making them the ideal general purpose bottle for solids. The amber color filters out ultraviolet wavelengths, making amber packers ideal for light sensitive solids. Amber packers are the classic vitamin bottles and are often used in pharmaceutical packaging. These packer bottles are also often used in chemical and environmental applications.
  • Attached polypropylene (PP) caps with F217 (PE foam) & PTFE liners are an economical alternative to Thermoset & Phenolic PTFE lined caps
  • F217 liners consist of 3 layers: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are very resilient and have strong resistance to chemicals and moisture to help keep bottle or jar contents free of odor and taste interference
Specifications
Capacity2 oz., 60mL
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorAmber
MaterialGlass
Packaging24/Cs.
Quantity24/Cs.
Closure ColorWhite
Liner MaterialPE Foam/PTFE
Neck Finish33-400
Certifications/ComplianceUSP Type III
Closure IncludedYes
MouthWide
ShapeRound
Product TypeWide Mouth Packer Bottle
Closure LinerPTFE/Polyethylene
Closure MaterialPolypropylene
Closure Size33-400 mm
Diameter (Metric)44 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePK