Traceable™ Bench Conductivity Meter
Traceable™ Bench Conductivity Meter
Actual product may vary
Fisherbrand
Traceable™ Bench Conductivity Meter
Traceable™ Bench Conductivity Meter measures conductivity, resistivity, dissolved solids, salinity, concentration, and temperature; includes certificate of traceability to standards provided by NIST™.
Have Questions?
Catalog NumberCalibration Points
FKS117642164
Catalog number FKS11764216
Price (SGD)
4,759.00
PK
Add to cart
Calibration Points:
4
Price (SGD)
4,759.00
PK
Add to cart
Traceable™ Bench Conductivity Meter measures conductivity, resistivity, dissolved solids, salinity, concentration, and temperature; includes certificate of traceability to standards provided by NIST™.
  • Features six modes — conductivity, resistivity, total dissolved solids, salinity, concentration and temperature readings
  • Ideal for ISO 9000 and government certification requirements
  • Autorange automatically selects most appropriate range for current reading unless feature is disabled
  • Controls allow operator to calibrate to solution standards; four calibration points ensure accuracy over entire range
Automatic or manual temperature compensation is defeatable
  • Automatic/manual temperature compensation with readings referenced to international standard of 25°C
  • Temperature compensation disable function fulfills USP-NF (United States Pharmacopeia, National Formulary, 645 Conductivity Measurement) requirements
Multiple applications plus connectivity
  • Measures conductivity in chromatography, biology, food, electronics and water analysis
  • Meets CAP, ASTM™, NCCLS, and ACS specifications in measuring reagent-grade water
  • Includes certificate to indicate instrument traceability to standards provided by the National Institute of Standards and Technology; calibrated by an ISO 17025 calibration laboratory accredited by A2LA
  • RS-232-C output allows readings to be downloaded to accessory datalogger or computer
Measurement Modes and Ranges
  • Conductivity Autoranging: 0.01 μS to 200 mS (in 5 ranges)
  • Resistance Autoranging: 0.001 to 20.00 megohms (in 2 ranges)
  • TDS Autoranging: 0.01 mg/L to 20.0 g/L (in 4 ranges)
  • Temperature: -30° to +130°C (-22° to +266°F)
  • Concentration: 0.000 to 9999 user units
  • Salinity: 2 to 42 ppt
  • Relative Accuracy: ±0.3%
Temperature Compensation
  • Types Automatic and manual, displays temperature °F/°C
  • Reference Temperature (conductivity): References to international standard 25°C (77°F)
Features
  • Electronic Calibration Supplied Internal (1000 °mhos and 0.001 megohm)
  • Data Output: RS-232-C, recorder jack
  • Display: 0.5 in. digit LCD
  • Net Weight: 1.5 lb. (0.7 kg)
  • Dimensions (L x W x H): 8.25 x 6 x 3.5 in. (21 x 15.2 x 8.9 cm)
Specifications
Accuracy (Conductivity)±0.3%
OutputsRS232
Includes115 V Power Adapter, 3 in. long (7.6 cm) Glass Probe, Receptacle to supply probe replatinizing current, 21 in. long (53.3 cm) Pivoting Probe Holder Arm, 9 V Battery
For Use With (Application)Conductivity
Measurement ModesConductivity, Resistivity, Dissolved Solid, Salinity, Concentration, Temperature
Calibration Points4
Range (Conductivity)0.01 μS to 200 mS
Range (Salinity)2 to 42 ppt
Range (TDS)0.01 mg/L to 20 g/L
Temperature Range (Metric)-30°C to +130°C
Temperature Range (English)-22°F to +266°F
Range (Resistivity)0.001 to 2 megohm
Display TypeLCD
Certifications/ComplianceCAP, ASTM, NCCLS, ACS
Weight (Metric)0.7 kg
Weight (English)1.5 lb.
Detection Range0.01 μS/cm to 200 mS/cm
Accuracy±0.3%
Height (English)8.25 in.
Height (Metric)20.95 cm
Depth (Metric)8.89 cm
Battery Type9 V
Voltage115 V
Unit SizePK