Microglass Fiber Filter Discs
Microglass Fiber Filter Discs
Actual product may vary
Fisherbrand
Microglass Fiber Filter Discs
Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filter Discs are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).
Have Questions?
Catalog NumberDiameter (Metric)
FKS1176408347 mm
Catalog number FKS11764083
Price (SGD)
138.00
PK
Add to cart
Diameter (Metric):
47 mm
Price (SGD)
138.00
PK
Add to cart
  • Contains no binder
  • Suitable for use at and up to 500°C
  • Not affected by humidity
  • Suitable for micro-filtration due to fine fibers
  • High-loading capacity
  • High chemical purity
  • made with demineralized water
  • High permeability to air
Applications:
  • Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
  • Sediments content in petroleum products
  • Ligand binding test
  • Air pollution monitoring
  • Membranes pre-filters
  • Radio-immuno tests
  • Suspended solids in water, smokes
  • Liquid scintillation counting
  • Cells collection
  • Grease content in dairy products
Specifications
Basis Weight75 g/m2
Filter TypeMicrofilter
FormatDisc
MaterialBorosilicate Glass
Pore Size0.7 μm
Filtration Time310 sec.
For Use With (Application)Air pollution control, separation of soot before gas analyzers, support weighing in the balance analyzes and dryers
Grade263
Quantity100 Pack
Surface TypeNon-hygroscopic
Temperature (Metric) Max500°C
Thickness (English)0.017 in.
Thickness (Metric)0.45 mm
TypeFilter
Diameter (Metric)47 mm
Unit SizePK