Square HDPE Bottles
Square HDPE Bottles
Actual product may vary
Fisherbrand
Square HDPE Bottles
Pre-dosed with sodium thiosulfate which inhibits the action of chlorine, bromine and ozone dissolved in water. Gosselin™ Square HDPE Bottles are manufactured from translucent HDPE with a graduated surface.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)IncludesPackaging
FKS11763104500 mL160 mm10 mg/L Sodium ThiosulfateIndividually Wrapped
Catalog number FKS11763104
Price (SGD)
668.00
PK
Add to cart
Capacity (Metric):
500 mL
Height (Metric):
160 mm
Includes:
10 mg/L Sodium Thiosulfate
Packaging:
Individually Wrapped
Price (SGD)
668.00
PK
Add to cart
  • Break resistant
  • Bottles are screw cap assembled (tamper evidence not engaged)
  • Sterilized by irradiation with peel-away sterilization certificate on the outer carton
Specifications
Capacity (Metric)500 mL
ColorTranslucent
Diameter (Metric) Inner Neck22 mm
Includes10 mg/L Sodium Thiosulfate
MaterialHDPE
Length (Metric)70mm
PackagingIndividually Wrapped
Quantity100/Carton
SterilitySterile
Width (Metric)70mm
Closure ColorRed
GraduationsYes
Certifications/ComplianceEC N°1935/2004, ISO 11137
Closure IncludedYes
ShapeSquare
Height (Metric)160 mm
Closure MaterialPolyethylene
Closure Type22 mm Red Tamper-evident Cap with Shaped Seal
Unit SizePK