LaMotte™ Secchi Turbidity Disk
LaMotte™ Secchi Turbidity Disk
Actual product may vary
Fisherbrand
LaMotte™ Secchi Turbidity Disk
Use this disk to determine turbidity or degree of visibility in natural water supplies. Cole-Parmer™ LaMotte™ Secchi Turbidity Disk features braided, stretch resistant lines marked at half and full meter intervals up to 20 meters.
Have Questions?
Catalog NumberProduct Type
FKS11762373Turbidity Disk
Catalog number FKS11762373
Price (SGD)
794.00
EA
Add to cart
Product Type:
Turbidity Disk
Price (SGD)
794.00
EA
Add to cart
Simple-to-use secchi disk is a weighted, 20-cm dia. black and white plate that sinks parallel to the water's surface. For best results use from the leeward side of a boat or dock between mid-morning and mid-afternoon.
Specifications
Product TypeTurbidity Disk
Quantity1 Ea.
Unit SizeEA