Sterileware™ Red Sterile Sampling Scoops
Sterileware™ Red Sterile Sampling Scoops
Actual product may vary
Fisherbrand
Sterileware™ Red Sterile Sampling Scoops
Perfect shape and size for taking samples, especially from wide-mouth containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FKS11761776250 mL100/Cs.
FIS03-990-228250 mL10/Pk.
FIS08-647-760250 mL100/Cs.
Catalog number FKS11761776
Price (SGD)
1,539.00
PK
Add to cart
Capacity (Metric):
250 mL
Quantity:
100/Cs.
Price (SGD)
1,539.00
PK
Add to cart
Perfect shape and size for taking samples, especially from wide-mouth containers.
  • Designed to hold various capacities, these one-piece scoops are molded of high-impact polystyrene.
  • 2 oz. and 4 oz. sampling scoops have a long handle that increases the distance between the hand and sample.
  • When placed on a flat surface they remain upright, can be used as weighing boats.
  • Colored scoop reduces the chance of being accidentally left behind in a sample; can serve as a code to avoid cross contamination and provide contrasting background for light colored samples.
  • Made from FDA grade materials.
  • Individually sealed and sterilized in a polyethylene bag ready for one time use and disposal.
  • Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization.
Specifications
Product TypeScoop
MaterialPolystyrene
AutoclavableNot Autoclavable
IncludesCertificate of sterilization
Capacity (English)8 oz.
Capacity (Metric)250 mL
DisposableSingle-use
SterilitySterile
ColorBright Red
Certifications/ComplianceFDA
For Use With (Application)Designed for taking samples, especially from wide-mouth containers, reduces the chance of residual sample staying in the scoop or the scoop being accidently left behind in a sample
For Use With (Equipment)Wide mouth containers
Quantity100/Cs.
Dimensions (L x W x H)7.5 x 2.12 x 1.93 in. (19 x 5.4 x 4.9 cm)
Height (English)1.93 in.
Height (English) Bowl1.93 in.
Height (Metric)4.9 cm
Height (Metric) Bowl4.9 cm
Length (English)7.5 in.
Length (English) Bowl5.31 in.
Length (English) Handle2.18 in.
Length (Metric)19 cm
Length (Metric) Bowl13.5 cm
Length (Metric) Handle5.6 cm
Width (English)2.12 in.
Width (English) Bowl2.12 in.
Width (Metric)5.4 cm
Width (Metric) Bowl5.4 cm
Unit SizePK