Microcon DNA Fast Flow Centrifugal Filter Unit with Ultracel Membrane
Microcon DNA Fast Flow Centrifugal Filter Unit with Ultracel Membrane
Actual product may vary
Fisherbrand
Microcon DNA Fast Flow Centrifugal Filter Unit with Ultracel Membrane
This 0.5 mL centrifugal filter is optimized for the concentration and recovery of genomic DNA with SDS buffer.
Have Questions?
Catalog NumberMWCO
FKS11760403100 kDa
Catalog number FKS11760403
Price (SGD)
-
MWCO:
100 kDa
Optimized for the concentration and recovery of genomic DNA.
Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes. With the low-binding Ultracel™ membrane, Microcon™ filters offer:•Typical recoveries of greater than 95%, even for dilute solutions •Reverse spin to maximize recovery, even in the smallest samples •Convenient storage of filtrate or concentrated sample in standard microfuge tube •Concentration factors up to 100X Microcon™ DNA Fast Flow FilterOptimized for the concentration and recovery of genomic DNA with SDS buffer. The low nonspecific binding characteristics of the membrane and the other device components, coupled with its medical-grade o-ring seal, allows the device to accommodate several wash steps with minimal sample loss.Features:•High recovery for small volumes with reverse spin (concentration factor less than 20X) •Low-binding Ultracel™ membrane •Fast processing Sample applications:•Recovery of genomic DNA for forensic applications •Concentration and desalting of nucleic acids (high recovery alternative to ethanol precipitation) •Removal of primers from amplified DNA •Easy filtrate recovery, exploiting the flat membrane design
Specifications
ColorBlue
DisposableSingle-use
Material (Membrane)Regenerated Cellulose
MWCO100 kDa
SterilityNon-sterile
Filtration Area0.32 cm2
Housing MaterialRegenerated Cellulose (RC)
TypeCentrifugal Filters
Volume (Metric)10 mL
Length (English)1.8 in.
Length (Metric)4.5 cm
Diameter (Metric)12.3 mm
Unit SizePK