Sterilin™ Homogenizer Bags, Steriblend
Sterilin™ Homogenizer Bags, Steriblend
Actual product may vary
Fisherbrand
Sterilin™ Homogenizer Bags, Steriblend
Made of food-grade heavy-gauge polyethylene, Thermo Scientific™ Sterilin™ Homogenizer Bags are ideal for homogenizing food samples before microbiological analysis.
Have Questions?
Catalog NumberDescriptionCapacity (Metric)Dimensions (L x W)No. per Case
FKS11758978Steriblend bag400 mL300 x 180mm500
Catalog number FKS11758978
Price (SGD)
390.00
PK
Add to cart
Description:
Steriblend bag
Capacity (Metric):
400 mL
Dimensions (L x W):
300 x 180mm
No. per Case:
500
Price (SGD)
390.00
PK
Add to cart
Made of food-grade heavy-gauge polyethylene, Thermo Scientific™ Sterilin™ Homogenizer Bags are ideal for homogenizing food samples before microbiological analysis.
  • Suitable for use in all leading homogenizer machines
  • Double-sealed for strength and reliability, ensuring safe processing of samples
  • Manufactured from food-grade heavy-guage polyethylene for durability
  • Sterilize by gamma irradiation
Ordering Information
  • Not available in all geographic regions
Specifications
DescriptionSteriblend bag
Capacity (Metric)400 mL
MaterialPolyethylene
No. per Case500
No. per Pack50
Dimensions (L x W)300 x 180mm
Length (Metric)300 mm
Width (Metric)180 mm
Unit SizePK