Stainless Steel Tall Form Container
Stainless Steel Tall Form Container
Actual product may vary
Fisherbrand
Stainless Steel Tall Form Container
Safely store and transport liquid gases with a container that includes a clamp to secure the lid to the body. Day Impex™ Stainless Steel Tall Form Container house a range of Dewar Flasks and are constructed of stainless steel for anti-corrosive properties.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Outer
FKS117563994500 mL182mm
FKS117463992 L142mm
Catalog number FKS11756399
Price (SGD)
1,445.00
EA
Add to cart
Capacity (Metric):
4500 mL
Diameter (Metric) Outer:
182mm
Price (SGD)
1,445.00
EA
Add to cart
  • Include clamp lid, vent, and handle
  • Vacuum insulated
  • Height dimesions include lid
  • Rust-free
  • Anti-corrosive
  • All hygienic
Specifications
Diameter (Metric) Outer182mm
For Use With (Application)For the transport of liquefied gases
Height (Metric)350mm
Height (Metric) Inner280mm
IncludesWith clamp lid attachment, vent and handle
Capacity (Metric)4500 mL
Depth (Metric) Interior280 mm
Diameter (Metric) Inner150 mm
MaterialStainless Steel
StyleTall Form
Unit SizeEA