Regulate flow without any leaking using these fittings. Cole-Parmer™ Plastic Luer Stopcocks and Manifolds are available with natural PVDF body and polypropylene insert, or all-black PVDF construction.
Have Questions?
Catalog number FKS11756138
Price (SGD)
403.00
EA
Add to cart
Price (SGD)
403.00
EA
Add to cart
Natural PVDF stopcocks available in 2- or 3-way configurations