High-Capacity Vacuum Pump, Gauge/Reg
High-Capacity Vacuum Pump, Gauge/Reg
Actual product may vary
Fisherbrand
High-Capacity Vacuum Pump, Gauge/Reg
Small, quiet source of vacuum or pressure
Have Questions?
Catalog number FKS11755539
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Small, quiet source of vacuum or pressure
  • Free-air capacities up to 2.2 cfm (62.3 L/min)
  • Shaded-pole, fan-cooled motor has thermal overload switch to protect your pump system
  • Different pump models have heads in parallel for higher capacity or series for greater vacuum
  • Select vacuum pumps with or without gauges, regulators, and valves
  • Runs dry
Specifications
Max. Vacuum24 Hg
Flow Rate28.3 L/min.
Pump TypeHigh Capacity Vacuum Pump
ApplicationsGast line of oil-less diaphragm air compressors and vacuum pumps
Maximum Pressure60 psi
Voltage220 V
Unit SizeEA