Check Order Status
Nickel/Plastic Bosshead
Actual product may vary
Fisherbrand
Nickel/Plastic Bosshead
For use with rods with a maximum diameter of 16mm.
Have Questions?
Catalog Number
Size
FKS11755386
16 mm
Catalog number
FKS11755386
Price (SGD)
55.00
EA
Add to cart
Size:
16 mm
Price (SGD)
55.00
EA
Add to cart
Supplied individually
With nickel plated steel screw
Specifications
Quantity
1 Ea.
Type
Bosshead Clamp
Material
Nickel, Plastic
Includes
Nickel plated brass screw Fitted blue Plastic headed screw
For Use With (Equipment)
Rods up to 16 mm dia.
Diameter (Metric)
16 mm
Size
16 mm