Polypropylene 2-place Slide Mailer
Polypropylene 2-place Slide Mailer
Actual product may vary
Fisherbrand
Polypropylene 2-place Slide Mailer
Store or ship two standard slides with confidence. Polypropylene 2-Place Slide Mailer has side-by-side slots in a sturdy polypropylene shell. Large, easy to secure snaps provide secure storage. Disposable.
Have Questions?
Catalog number FKS11755088
Price (SGD)
63.00
PK
Add to cart
Price (SGD)
63.00
PK
Add to cart
  • Protectors hold two standard slides: 1 x 3 in. or 25 x 75 mm
  • Large, easy snap lids ensure slides stay securely in the mailer
  • Slides lay flat in separate channels to avoid slide contact or shifting
  • 25 disposable mailers per pack
Specifications
TypeSlide Mailer
Slide Capacity2
ColorNatural
Device HoldsSlides
For Use With (Equipment)Standard Microscope Slides
Height (English)0.2 in.
Height (Metric)5 mm
Length (English)3.7 in.
Length (Metric)93 mm
MaterialPolypropylene
Width (English)2.7 in.
Width (Metric)68 mm
Unit SizePK