Amber Soda Glass Wide Mouth Packer Bottles with White Polypropylene/Polyvinyl Lined Cap
Amber Soda Glass Wide Mouth Packer Bottles with White Polypropylene/Polyvinyl Lined Cap
Actual product may vary
Fisherbrand
Amber Soda Glass Wide Mouth Packer Bottles with White Polypropylene/Polyvinyl Lined Cap
Ideal for storage of dry powders.
Have Questions?
Catalog NumberCapacity (Metric)Quantity
FKS1175431930 mL24/Cs.
Catalog number FKS11754319
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
30 mL
Quantity:
24/Cs.
  • Wide mouth for bottles allows easy accessibility for removal of contents
  • Custom cleaned containers and caps are free of visible particulates done using deionised water
  • Certified clean containers are cleaned as per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
Specifications
Capacity1 oz., 30mL
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorAmber
Diameter (Metric) Inner Neck15.1mm
IncludesCertificate of analysis
MaterialSoda Lime Glass
Packaging24/Cs.
Quantity24/Cs.
Closure ColorWhite
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)65 mm
Product TypeWide Mouth Packer Bottle
Closure LinerPolyvinyl
Closure MaterialPolypropylene
Closure Size28-400 mm
Diameter (Metric)37 mm
Closure TypeScrew Cap
Unit SizePK