Digital Auto-Range Multimeter
Digital Auto-Range Multimeter
Actual product may vary
Fisherbrand
Digital Auto-Range Multimeter
Designed specifically for scientific equipment, this unit automatically selects proper range when testing electrical functions of instruments and their power sources
Have Questions?
Catalog number FKS11754276
Price (SGD)
-
Designed specifically for scientific equipment, this unit automatically selects proper range when testing electrical functions of instruments and their power sources
  • Unit ranges are from 1mV to 400V AC/DC and 0.1 to 2,000,000ohms with accuracy to 2 units (2%)
  • Sampling rate is twice per second
  • Features audible beep for checking continuity
Specifications
Accuracy2%
Certifications/ComplianceNIST
Dimensions (L x W x H)2.125 x 4.25 x .5 in.
For Use With (Application)For scientific equipment, this unit automatically selects proper range when testing electrical functions of instruments and their power sources
Height (English)0.5 in.
Height (Metric)1.3 cm
IncludesCase, batteries, probes and instructions
Length (English)2.12 in.
Length (Metric)5.4 cm
Weight (English)0.17 lb.
Weight (Metric)0.08 kg
Width (English)4.25 in.
Width (Metric)10.8 cm
Product TypeMultimeter, Digital Auto Range
Voltage1 mV/400 V
Unit SizeEA