Check Order Status
Class A Brown Graduated USP Measuring Pipettes
Actual product may vary
Fisherbrand
Class A Brown Graduated USP Measuring Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs.
Have Questions?
Change view
Catalog Number
Volume (Metric)
FKS11751526
50 mL
FKS11741526
25 mL
FKS11731536
100 mL
3 Options
Catalog number
FKS11751526
Price (SGD)
341.00
PK
Add to cart
Volume (Metric):
50 mL
Price (SGD)
341.00
PK
Add to cart
With dated batch identification
Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
With volume mark
Brown graduation
Specifications
Class
A
Material
AR-GLAS
For Use With (Application)
Volumetric accuracy in pipetting
Graduated
Yes
Graduation Color
Brown
Tolerance
0.05 mL
Volume (Metric)
50 mL
Length (Metric)
540 ±10 mm