Blood Collection Tray
Blood Collection Tray
Actual product may vary
Fisherbrand
Blood Collection Tray
Designed with phlebotomists in mind. Fisherbrand™ Blood Collection Tray was developed after extensive research in clinics and hospitals. This lightweight solution for organizing and transporting tubes and smaller items includes a sturdy plastic tray, 36-well tube rack and disposable inserts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
FKS1175063416 mm tube rack (Style B)
FIS22-267-27013 mm tube rack (Style A)
FKS1174063413 mm tube rack (Style A)
FIS22-267-27116 mm tube rack (Style B)
Catalog number FKS11750634
Price (SGD)
450.00
EA
Add to cart
For Use With (Equipment):
16 mm tube rack (Style B)
Price (SGD)
450.00
EA
Add to cart
Designed with phlebotomists in mind. Fisherbrand™ Blood Collection Tray was developed after extensive research in clinics and hospitals. This lightweight solution for organizing and transporting tubes and smaller items includes a sturdy plastic tray, 36-well tube rack and disposable inserts.
  • Lightweight, washable ABS tray
  • Tray features a supply drawer, side pockets, and a ratcheting handle with soft rubber grip that rotates out of the way for easy access
  • Tray includes a Heathrow Onerack™ tube rack and two disposable polystyrene inserts
  • Additional inserts sold in packs of 4 in two different configurations
Specifications
ColorBlue
Dimensions (L x W x H)429 x 265 x 172 mm
For Use With (Application)Blood collection
For Use With (Equipment)16 mm tube rack (Style B)
Includes2 inserts, 16 mm test tube rack, supply drawer and side pockets, and ratcheting handle
No. of Wells36 (tube rack)
MaterialABS
Product TypeBlood Collection Tray
Unit SizeEA