Traceable™ Flashing LED Big-Digit Dual Channel Timer
Traceable™ Flashing LED Big-Digit Dual Channel Timer
Actual product may vary
Traceable™ Flashing LED Big-Digit Dual Channel Timer
Traceable™ Flashing LED Big-Digit Dual Channel Timer
Fisherbrand
Traceable™ Flashing LED Big-Digit Dual Channel Timer
Times two different activities simultaneously
Have Questions?
Catalog number FKS11749795
Price (SGD)
133.00
EA
Add to cart
Price (SGD)
133.00
EA
Add to cart
Times two different activities simultaneously. Fisher Scientific™ Traceable™ Flashing LED Big-Digit Dual Channel Timer has a large, 0.63 in.-high LCD digits that can be seen from 20 feet away.
  • Time each channel counts separately: 99 hr. 59 min. 59 sec. down to 1 sec.
  • Each channel has a distinct alarm and visual indicator when zero is reached
  • Green light flashes during countdown; red after zero is reached and alarm is sounding
  • Audible alarms have three settings: high, low and muted (silent)
  • Alarm may be turned off or silences automatically after 1 min.
  • For repetitive timing, microcomputer chip memory permits two countdown times
  • For normal timing, memory recalls last countdown time set
  • Magnetic back, flip-open easel and opening for lanyard for easy placement of timer
  • Accuracy: 0.01%
  • Resolution: 1 sec.
  • Specifications
    Product TypeHandheld/Benchtop Timer
    Time Range99 hr. 59 min. 59 sec.
    AlarmsAudible/Visible
    Accuracy0.0001
    Resolution1 sec.
    DisplayLED
    Dimensions (L x W x H)3.5 x 0.63 x 3 in. (8.9 x 1.6 x 7.6 cm)
    Length (English)3.5 in.
    Length (Metric)8.9 cm
    Width (English)0.63 in.
    Width (Metric)1.6 cm
    Height (English)3 in.
    Height (English) Display0.62 in.
    Height (Metric)7.6 cm
    Height (Metric) Display1.57 cm
    Weight (English)3 oz.
    Weight (Metric)85 g
    IncludesBattery, magnetic back, flip-open stand/clip, opening for lanyard, Traceable™ Certificate.
    Certifications/ComplianceISO 17025, A2LA, NIST
    Unit SizeEA