Disposable Aluminum Dishes with Fluted Sides
Disposable Aluminum Dishes with Fluted Sides
Actual product may vary
Fisherbrand
Disposable Aluminum Dishes with Fluted Sides
Useful as evaporating and general utility dishes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Inner TopDiameter (Metric) Inner Base
FKS1174941875 mL62 mm46 mm
FIS08-732-10520 mL43 mm40 mm
FIS08-732-10660 mL64 mm59 mm
FIS08-732-10775 mL62 mm46 mm
FIS08-732-109500 mL180 mm137 mm
FIS08-732-108200 mL109 mm77 mm
Catalog number FKS11749418
Price (SGD)
212.00
PK
Add to cart
Capacity (Metric):
75 mL
Diameter (Metric) Inner Top:
62 mm
Diameter (Metric) Inner Base:
46 mm
Price (SGD)
212.00
PK
Add to cart
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides are inexpensive enough for single use; useful as evaporating and general utility dishes
Larger sizes are ideal for food processing applications.
  • Made of lightweight aluminum
  • Flat bottoms
  • Fluted or slightly fluted sides
  • Flanged rims
Specifications
Bottom ShapeFlat
Capacity (Metric)75 mL
DisposableSingle-use
MaterialAluminum
ShapeRound
TypeWeighing Dish
For Use With (Application)Useful as evaporating and general utility dishes
Diameter (Metric) Inner Base46 mm
Diameter (Metric) Inner Top62 mm
Diameter (Metric)46 mm (Inner Base), 62 mm (Inner Top)
Unit SizePK