A-Shaped Retort Stand Base
A-Shaped Retort Stand Base
Actual product may vary
A-Shaped Retort Stand Base
A-Shaped Retort Stand Base
A-Shaped Retort Stand Base
A-Shaped Retort Stand Base
An excellent option for supporting retort stands in the laboratory. R&L Enterprises™ A-Shaped Retort Stand Bases are manufactured from cast iron with a chemically resistant blue acrylic paint.
Have Questions?
Catalog NumberWeight (Metric)
FKS117493272.5 kg
Catalog number FKS11749327
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Weight (Metric):
2.5 kg
  • Supplied without rod
  • Supplied with thumbscrew
  • Mounting rod hole is dual purpose and will accept both plain rods and M10 screwed rods of 12.7mm in diameter
Specifications
MaterialCast Iron
Product TypeTripod Stand
For Use With (Application)Fixings For Rods Or Tubes
Weight (Metric)2.5 kg
Unit SizeEA