Sterilin™ 30 to 140mm Petri Dishes
Sterilin™ 30 to 140mm Petri Dishes
Actual product may vary
Sterilin™ 30 to 140mm Petri Dishes
Sterilin™ 30 to 140mm Petri Dishes
Fisherbrand
Sterilin™ 30 to 140mm Petri Dishes
Save incubator space with Thermo Scientific™ Sterilin™ 30 to 140mm Petri Dishes. These small dishes are suitable for most applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)Vent
FKS1174924255 mm12 mmNo
FKS1132928335 mm11 mm3
FKS1136927360 mm15 mm1
Catalog number FKS11749242
Price (SGD)
2,128.00
PK
Add to cart
Diameter (Metric):
55 mm
Height (Metric):
12 mm
Vent:
No
Price (SGD)
2,128.00
PK
Add to cart
Save incubator space with Thermo Scientific™ Sterilin™ 30 to 140mm Petri Dishes. These small dishes are suitable for most applications.
NOT AVAILABLE IN AMERICA
  • 50mm diameter dish (catalog number 124) is over 20mm deep and designed for use with liquid media to aid handling
  • 55mm dish accommodates 47mm membrane filters making it suitable for water testing
  • 140mm dish is produced and tested in accordance with the EN ISO 24998 Petri dish standard which includes stringent dimensional controls
Specifications
ShapeRound
SterilityAseptically Manufactured
TypePetri Dish
No. per Case1620
No. per Pack15
VentNo
Height (Metric)12 mm
Height (English)12 in.
Diameter (Metric)55 mm
Unit SizePK