Double Burette Clamp
Double Burette Clamp
Actual product may vary
Fisherbrand
Double Burette Clamp
Securely holds burettes in place during the construction of laboratory retort stands. R&L Enterprises™ Double Burette Clamps are suitable for burettes up to 100mL capacity.
Have Questions?
Catalog NumberFor Use With (Equipment)
FKS11749027For use up to 100mL burette
Catalog number FKS11749027
Price (SGD)
137.00
EA
Add to cart
For Use With (Equipment):
For use up to 100mL burette
Price (SGD)
137.00
EA
Add to cart
  • Diecast
  • Without rod
  • With a single plastic headed thumbscrew to secure the clamp to retort rods, vertically or horizontally
Specifications
Quantity1 Ea.
TypeDouble Clamp
Includeswith a single Plastic headed thumbscrew to secure the clamp to retort rods
For Use With (Equipment)For use up to 100mL burette
Unit SizeEA