Wheaton™ Tapered Tissue Grinders With PTFE Pestles
Wheaton™ Tapered Tissue Grinders With PTFE Pestles
Actual product may vary
Fisherbrand
Wheaton™ Tapered Tissue Grinders With PTFE Pestles
Tapered tissue grinder with a PTFE pestle for soft tissues
Have Questions?
Catalog NumberCapacityLength (English) PestleLength (English) TubeTube Diameter (English) Outside
FKS117489390.5 oz., 15mL9.4 in.4.48 in.0.7 in.
Catalog number FKS11748939
Price (SGD)
875.00
EA
Add to cart
Capacity:
0.5 oz., 15mL
Length (English) Pestle:
9.4 in.
Length (English) Tube:
4.48 in.
Tube Diameter (English) Outside:
0.7 in.
Price (SGD)
875.00
EA
Add to cart
Stainless steel rod is 0.25 in. (6.3mm) in diameter and long enough to fit into the chuck of the Wheaton Overhead Stirrer.
  • PTFE pestle with stainless steel rod fits power homogenizer
  • Homogenizes soft tissue such as brain
  • Mortar made from low-extractable borosilicate glass
  • Interchangeable components takes the hassle out of matching parts
  • Autoclavable
Specifications
Capacity (English)0.5 oz.
Capacity (Metric)15 mL
MaterialBorosilicate Glass, PTFE
Product TypeTissue Grinder
IncludesWith Teflon Stamp
Length (English) Pestle9.4 in.
Length (Metric) Pestle240mm
Length (English) Tube4.48 in.
Length (Metric) Tube114mm
Tube Diameter (English) Outside0.7 in.
Tube Diameter (Metric) Outside18mm
Capacity0.5 oz., 15mL
Unit SizeEA