Fritted Glass Support for 47mm Filtration Assembly with Standard Taper Joint Connection
Fritted Glass Support for 47mm Filtration Assembly with Standard Taper Joint Connection
Actual product may vary
Fisherbrand
Fritted Glass Support for 47mm Filtration Assembly with Standard Taper Joint Connection
Recommended for general filtration. Wheaton™ Fritted Glass Support for 47mm Filtration Assembly with Standard Taper Joint Connection is manufactured from low extractable borosilicate glass.
Have Questions?
Catalog NumberType
FKS11748799Vaccum Filter
Catalog number FKS11748799
Price (SGD)
2,734.00
PK
Add to cart
Type:
Vaccum Filter
Price (SGD)
2,734.00
PK
Add to cart
  • Includes a coarse porosity (40-60μm) fritted glass support base, graduated funnel, anodized aluminum clamp and 1L filter flask
  • Glass components manufactured from Wheaton™ 33 glass which conforms to USP Type 1 and ASTM E Type I, Class A requirements
Specifications
For Use With (Application)Routine Filtration Of Organic Solvents, Corrosive Liquids And The Removal Of Particulates From HPLC Solvents
TypeVaccum Filter
Volume (Metric)500 mL
Unit SizePK