Check Order Status
Polystyrene Beakers
Actual product may vary
Fisherbrand
Polystyrene Beakers
Ideal for use as sample containers
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS11748549
50 mL
FIS08-732-119
5 mL
FIS08-732-121
10 mL
FIS08-732-122
50 mL
4 Options
Catalog number
FKS11748549
Price (SGD)
92.00
PK
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
92.00
PK
Add to cart
Ideal for use as sample containers
20mL size is useful in flame photometry
5mL size can be used in atomic absorption spectroscopy and flame photometry
Specifications
Diameter (Metric) Base
31 mm
Diameter (Metric)Top
61 mm
Capacity (Metric)
50 mL
Material
Polystyrene
Type
Beaker
Disposable
Single-use