Stainless Steel Micro Spoons
Stainless Steel Micro Spoons
Actual product may vary
Fisherbrand
Stainless Steel Micro Spoons
Designed using high quality components these micro spoons feature a natural quality and bright finish.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS11746607100 mm
FKS11593482150 mm
Catalog number FKS11746607
Price (SGD)
83.00
Pack
Add to cart
Length (Metric):
100 mm
Price (SGD)
83.00
Pack
Add to cart
  • Designed using high quality components these micro spoons feature a natural quality and bright finish
  • Laboratory Micro Spoon, stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Length = 100mm long
  • Spoon measurement 8mm wide x 6mm long x 1.5mm deep
  • Centre barrel diameter 1.60mm - 'micro'
  • Spoon capacity [liquid] = 0.2ml
  • Supplied in standard packs of 10
Specifications
Capacity (Metric)0.2 mL
Product TypeSpoon
FinishBright
MaterialStainless Steel
Length (Metric)100 mm
Unit SizePack