Traceable™ Big-Digit Timer/Stopwatch
Traceable™ Big-Digit Timer/Stopwatch
Actual product may vary
Fisherbrand
Traceable™ Big-Digit Timer/Stopwatch
Set four separate channels from 99 hours, 59 minutes, 59 seconds to one second. Fisherbrand™ Traceable™ Big Digit Timer/Stopwatch sounds an alarm when zero is reached and then starts counting up.
Have Questions?
Catalog number FKS11745863
Price (SGD)
126.00
EA
Add to cart
Price (SGD)
126.00
EA
Add to cart
  • Giant 0.75 in. (19mm) display for greater readability
  • Features include count down, count up (stopwatch), time-out and time of day
  • When zero is reached, alarm sounds and timer starts counting up
  • All four channels may be used simultaneously in any combination of count down/count up
  • Equivalent battery replacements are: RAYOVAC RW42, DURACELL D357, and EVEREADY 357.
Specifications
Product Type4-channel Alarm Timer
Time Range99 hr. 59 min. 59 sec.
Accuracy0.0001
Resolution1 sec.
DisplayLCD
Dimensions (L x W x H)2.75 x 2.5 x 0.5 in. (7 x 6.4 x 1.3 cm)
Length (English)2.75 in.
Length (Metric)7 cm
Width (English)2.5 in.
Width (Metric)6.4 cm
Height (English)0.5 in.
Height (English) Display0.75 in.
Height (Metric)1.3 cm
Height (Metric) Display1.9 cm
Weight (English)2 oz.
Weight (Metric)56.69 g
IncludesSilver Oxide battery
For Use With (Application)High display that permits viewing from across the lab
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEA