Check Order Status
Bossheads
Actual product may vary
Fisherbrand
Bossheads
For securely keeping clamps in place on laboratory scaffolding. R&L Enterprises™ Bossheads come with offset jaws for greater strength.
Have Questions?
Catalog Number
Includes
FKS11745386
Blue Plastic headed screw
Catalog number
FKS11745386
Price (SGD)
29.00
Each
Add to cart
Includes:
Blue Plastic headed screw
Price (SGD)
29.00
Each
Add to cart
Aluminum alloy with clamping screws
For use with rods up to 16mm in diameter
Specifications
Quantity
1 Ea.
Type
Bosshead Clamp
Includes
Blue Plastic headed screw
For Use With (Equipment)
For Rods up to 16mm dia.
Diameter (Metric)
16 mm