Nylon Membrane Filter
Nylon Membrane Filter
Actual product may vary
Fisherbrand
Nylon Membrane Filter
Naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.
Have Questions?
Catalog NumberPore SizeDiameter (Metric)
FKS117453150.22 μm47 mm
Catalog number FKS11745315
Price (SGD)
331.00
PK
Add to cart
Pore Size:
0.22 μm
Diameter (Metric):
47 mm
Price (SGD)
331.00
PK
Add to cart
  • Naturally hydrophilic membrane
  • Designed to wet out evenly and retain its superior strength during use in general filtration or medical assays
  • Flexible, durable and tear resistant
  • Autoclaved at 135°C
  • Effective chemical compatibility with aqueous and alcoholic solutions and solvents
Specifications
AutoclavableAutoclavable
Bubble Point3.4bar
ColorWhite
Filter TypeMicrofilter
MaterialNylon
Pore Size0.22 μm
SterilityNon-sterile
Flow Rate9.9mL/min./cm2
For Use With (Application)Sterilizing filtration of aqueous solutions, organic solvents, Bacterial and particles removal (blood Glucose, Serum Cholesterol, drugs)
Gas Flow Rate1.7L/min./cm2
HydrophobicityHydrophilic
Quantity100 Pack
TypePlain Disc
WettabilityYes
Diameter (Metric)47 mm
Unit SizePK