Permanent Markers
Permanent Markers
Actual product may vary
Fisherbrand
Permanent Markers
Write on all surfaces
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantity
FKS11744386Black6 Pk.
CTL13-383Assorted12 Pk.
FIS13-383Assorted12 Pk.
CTL13-383-CBlack6 Pk.
FIS13-383CBlack6 Pk.
FIS13-383FGreen6 Pk.
Catalog number FKS11744386
Price (SGD)
70.00
PK
Add to cart
Color:
Black
Quantity:
6 Pk.
Price (SGD)
70.00
PK
Add to cart
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
  • Produce precise, ultrathin lines 0.3mm wide
  • Waterproof and fadeproof, permanent ink dries almost instantly
  • An airtight ring seal cap prevents dry-out and ensures instant writing even after lengthy storage
  • Unique stainless-steel long leg allows precise writing in a confined space
Specifications
Product TypePermanent Pen
Tip Style0.3 mm Ultra Fine
ColorBlack
DescriptionNot chemical resistant
MaterialStainless Steel
For Use With (Application)Perfect marking pens for lab samples, microscope slides, photographic slides, glass and plastic beakers, and plastic sample bags
Quantity6 Pk.
Width (Metric) Line0.3 mm
Unit SizePK